Product Details

SNP ID
rs138815958
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:47994357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTGCACTGGAGATGTTTCCTGA[C/T]GGAGACACCTTCTCTGAGATGTGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SUN3 PubMed Links

Gene Details

Gene
SUN3
Gene Name
Sad1 and UNC84 domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030019.1 906 Silent Mutation CCC,CCT P273P NP_001025190.1
NM_001284350.1 906 Silent Mutation CCC,CCT P261P NP_001271279.1
NM_152782.3 906 Silent Mutation CCC,CCT P273P NP_689995.3
XM_017011930.1 906 Silent Mutation CCC,CCT P273P XP_016867419.1

View Full Product Details