Product Details

SNP ID
rs139051155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99928874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATTGCTCTTTGGTTTCCACCAC[G/T]GGGAGGCTATCGGTTGCTTTCTTGT
Phenotype
MIM: 611925
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GJC3 PubMed Links

Gene Details

Gene
GJC3
Gene Name
gap junction protein gamma 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181538.2 747 Silent Mutation CCA,CCC P249P NP_853516.1
Gene
LOC101927610
Gene Name
uncharacterized LOC101927610
There are no transcripts associated with this gene.

Gene
TRIM4
Gene Name
tripartite motif containing 4
There are no transcripts associated with this gene.

View Full Product Details