Product Details

SNP ID
rs139708297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:140674121 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCTTGGAAATGGCCGGAAACCTC[C/G]AGAAAATCTCGCAGACCAGTCGTTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADCK2 PubMed Links

Gene Details

Gene
ADCK2
Gene Name
aarF domain containing kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052853.3 1194 Missense Mutation CCA,CGA P264R NP_443085.2
XM_006716170.3 1194 Missense Mutation CCA,CGA P264R XP_006716233.1
XM_011516675.2 1194 Missense Mutation CCA,CGA P264R XP_011514977.1

View Full Product Details