Product Details
- SNP ID
-
rs139741634
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:100171263 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTCAGGGATGCAGGGGTCTCACCC[G/T]CGTCCTGCCCGTCCAGGTCGTGTCC
- Phenotype
-
MIM: 608235
MIM: 608489
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GAL3ST4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75779608] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GAL3ST4
- Gene Name
- galactose-3-O-sulfotransferase 4
There are no transcripts associated with this gene.
- Gene
- GPC2
- Gene Name
- glypican 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152742.2 |
1711 |
Missense Mutation |
GAG,GCG |
E495A |
NP_689955.1 |
- Gene
- STAG3
- Gene Name
- stromal antigen 3
There are no transcripts associated with this gene.
View Full Product Details