Product Details

SNP ID
rs140792992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101206362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCGTGGTAGTGGGTGAGGCGGCC[A/G]GGGTGCAGGAGTGCCCAGCCCTTCC
Phenotype
MIM: 610184 MIM: 603066
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MOGAT3 PubMed Links

Gene Details

Gene
MOGAT3
Gene Name
monoacylglycerol O-acyltransferase 3
There are no transcripts associated with this gene.

Gene
PLOD3
Gene Name
procollagen-lysine,2-oxoglutarate 5-dioxygenase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001084.4 2592 Silent Mutation CCC,CCT P712P NP_001075.1

View Full Product Details