Product Details

SNP ID
rs140905443
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:115936480 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTCCACATCTTACACATCAGAC[A/G]GTCAGTAAATACCTAAAGTGTTTGA
Phenotype
MIM: 604732
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TFEC PubMed Links

Gene Details

Gene
TFEC
Gene Name
transcription factor EC
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018058.2 5570 UTR 3 NP_001018068.1
NM_001244583.1 5570 UTR 3 NP_001231512.1
NM_012252.3 5570 UTR 3 NP_036384.1
XM_011515963.1 5570 UTR 3 XP_011514265.1
XM_011515964.2 5570 UTR 3 XP_011514266.1
XM_011515965.2 5570 UTR 3 XP_011514267.1
XM_011515966.2 5570 UTR 3 XP_011514268.1
XM_011515969.1 5570 UTR 3 XP_011514271.1
XM_011515970.1 5570 UTR 3 XP_011514272.1
XM_011515972.2 5570 UTR 3 XP_011514274.1
XM_017011874.1 5570 UTR 3 XP_016867363.1
XM_017011875.1 5570 UTR 3 XP_016867364.1
XM_017011876.1 5570 UTR 3 XP_016867365.1
XM_017011877.1 5570 UTR 3 XP_016867366.1
XM_017011878.1 5570 UTR 3 XP_016867367.1

View Full Product Details