Product Details
- SNP ID
-
rs140946542
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:87402240 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTCTGCATTCTGGATGGTGGACAG[A/G]CGGTGAGCAATCACAATGCAGGTGC
- Phenotype
-
MIM: 171060
MIM: 606090
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ABCB4
PubMed Links
Gene Details
- Gene
- ABCB4
- Gene Name
- ATP binding cassette subfamily B member 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000443.3 |
3654 |
Silent Mutation |
CGC,CGT |
R1232R |
NP_000434.1 |
NM_018849.2 |
3654 |
Silent Mutation |
CGC,CGT |
R1239R |
NP_061337.1 |
NM_018850.2 |
3654 |
Silent Mutation |
CGC,CGT |
R1185R |
NP_061338.1 |
XM_011516308.2 |
3654 |
Silent Mutation |
CGC,CGT |
R1239R |
XP_011514610.2 |
XM_011516309.2 |
3654 |
Silent Mutation |
CGC,CGT |
R1232R |
XP_011514611.2 |
XM_011516310.2 |
3654 |
Silent Mutation |
CGC,CGT |
R1204R |
XP_011514612.2 |
XM_011516311.2 |
3654 |
Silent Mutation |
CGC,CGT |
R1196R |
XP_011514613.2 |
XM_011516312.2 |
3654 |
Silent Mutation |
CGC,CGT |
R1192R |
XP_011514614.2 |
XM_011516313.2 |
3654 |
Silent Mutation |
CGC,CGT |
R1185R |
XP_011514615.1 |
XM_011516315.2 |
3654 |
Silent Mutation |
CGC,CGT |
R1019R |
XP_011514617.2 |
XM_017012323.1 |
3654 |
Silent Mutation |
CGC,CGT |
R1239R |
XP_016867812.1 |
- Gene
- CROT
- Gene Name
- carnitine O-octanoyltransferase
There are no transcripts associated with this gene.
View Full Product Details