Product Details

SNP ID
rs141383805
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:157337816 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAACTCTTACCTAAGGAAGGTCA[C/G]TGACACACGCAGTGTAAACTGCAAA
Phenotype
MIM: 611332
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNAJB6 PubMed Links

Gene Details

Gene
DNAJB6
Gene Name
DnaJ heat shock protein family (Hsp40) member B6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005494.2 Intron NP_005485.1
NM_058246.3 Intron NP_490647.1
XM_005249515.2 Intron XP_005249572.1
XM_005249516.2 Intron XP_005249573.1
XM_006715823.1 Intron XP_006715886.1
XM_011515704.1 Intron XP_011514006.1
XM_017011633.1 Intron XP_016867122.1

View Full Product Details