Product Details

SNP ID
rs141401904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149199150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGAGTCGCACCTCAAACCAGGCT[A/G]CCTGGAGTTGCTTTCAAGTCGTAGT
Phenotype
MIM: 603397
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF282 PubMed Links

Gene Details

Gene
ZNF282
Gene Name
zinc finger protein 282
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303481.1 Intron NP_001290410.1
NM_003575.3 Intron NP_003566.1
XM_006716151.3 Intron XP_006716214.1

View Full Product Details