Product Details

SNP ID
rs141446586
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:116215965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTCCCATTCTTCATGCAAAAAC[A/G]TTTCTTATGAAGCTGAGAAGCTGTG
Phenotype
MIM: 606085
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TES PubMed Links

Gene Details

Gene
TES
Gene Name
testin LIM domain protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015641.3 Intron NP_056456.1
NM_152829.2 Intron NP_690042.1
XM_005250258.1 Intron XP_005250315.1

View Full Product Details