Product Details

SNP ID
rs142331989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:122304225 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCACGAAGGGGATCATGCAGGG[A/G]ATCGACGAGTTGAGATGCAGAGAGT
Phenotype
MIM: 613301
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FEZF1 PubMed Links

Gene Details

Gene
FEZF1
Gene Name
FEZ family zinc finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024613.3 1958 Silent Mutation ATC,ATT I71I NP_001019784.2
NM_001160264.2 1958 Silent Mutation ATC,ATT I71I NP_001153736.1
XM_005250337.3 1958 Silent Mutation ATC,ATT I71I XP_005250394.1
XM_011516202.2 1958 Silent Mutation ATC,ATT I71I XP_011514504.1
Gene
FEZF1-AS1
Gene Name
FEZF1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details