Product Details

SNP ID
rs142480920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:121873541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCCTCGCTTGCATTCAGCTCCT[C/T]TGTGTTTGCCGCCTGGGTGAGTGAG
Phenotype
MIM: 176891
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTPRZ1 PubMed Links

Gene Details

Gene
PTPRZ1
Gene Name
protein tyrosine phosphatase, receptor type Z1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206838.1 437 Silent Mutation CTC,CTT L14L NP_001193767.1
NM_001206839.1 437 Silent Mutation CTC,CTT L14L NP_001193768.1
NM_002851.2 437 Silent Mutation CTC,CTT L14L NP_002842.2
XM_005250519.1 437 Silent Mutation CTC,CTT L14L XP_005250576.1
XM_017012477.1 437 UTR 5 XP_016867966.1

View Full Product Details