Product Details

SNP ID
rs142949198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143382873 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTCCAGTACCAAGCCTGCAGCC[G/T]GGGGCACAGCACCCCTGCCTCCTTG
Phenotype
MIM: 179610 MIM: 602002
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EPHA1 PubMed Links

Gene Details

Gene
EPHA1
Gene Name
EPH receptor A1
There are no transcripts associated with this gene.

Gene
FAM131B
Gene Name
family with sequence similarity 131 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031690.2 627 Intron NP_001026860.2
NM_001278297.1 627 Intron NP_001265226.1
NM_014690.4 627 Intron NP_055505.3
XM_005250073.3 627 Intron XP_005250130.1
XM_006716186.2 627 Intron XP_006716249.1
Gene
LOC100507507
Gene Name
uncharacterized LOC100507507
There are no transcripts associated with this gene.

Gene
MIR6892
Gene Name
microRNA 6892
There are no transcripts associated with this gene.

Gene
ZYX
Gene Name
zyxin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010972.1 627 Missense Mutation GGG,TGG G192W NP_001010972.1
NM_003461.4 627 Missense Mutation GGG,TGG G192W NP_003452.1
XM_011516569.2 627 Missense Mutation GGG,TGG G161W XP_011514871.2
XM_017012587.1 627 Missense Mutation GGG,TGG G251W XP_016868076.1

View Full Product Details