Product Details

SNP ID
rs143988787
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73570101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGAAGCTATCCCAGTCTCCCATC[C/G]TTGCAAAACTGCTGCTTAGTACTCA
Phenotype
MIM: 605842
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TBL2 PubMed Links

Gene Details

Gene
TBL2
Gene Name
transducin (beta)-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012453.2 1754 UTR 3 NP_036585.1
XM_006715923.3 1754 UTR 3 XP_006715986.1
XM_011516027.2 1754 UTR 3 XP_011514329.1

View Full Product Details