Product Details

SNP ID
rs144875801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6501808 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGACCATTGCAAACTTGTCCTC[A/G]CTAGAGGGGGAAATGCTCTGGCAGG
Phenotype
MIM: 610639
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRID2IP PubMed Links

Gene Details

Gene
GRID2IP
Gene Name
Grid2 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145118.1 3372 Silent Mutation AGC,AGT S1124S NP_001138590.1
XM_011515404.2 3372 Silent Mutation AGC,AGT S940S XP_011513706.1

View Full Product Details