Product Details

SNP ID
rs145152108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:103075779 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAGGTCAATCTCTGCTTGCAGGT[G/T]CCCTGGAAGAAGGGACGTCAGAGGG
Phenotype
MIM: 611864 MIM: 609080
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARMC10 PubMed Links

Gene Details

Gene
ARMC10
Gene Name
armadillo repeat containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161009.2 534 Intron NP_001154481.1
NM_001161010.2 534 Missense Mutation GCC,TCC A48S NP_001154482.1
NM_001161011.2 534 Intron NP_001154483.1
NM_001161012.2 534 Intron NP_001154484.1
NM_001161013.2 534 Intron NP_001154485.1
NM_031905.4 534 Missense Mutation GCC,TCC A48S NP_114111.2
XM_005250628.2 534 Missense Mutation GCC,TCC A48S XP_005250685.1
XM_011516601.2 534 Intron XP_011514903.1
XM_011516602.2 534 Intron XP_011514904.1
XM_017012684.1 534 Missense Mutation GCC,TCC A48S XP_016868173.1
XM_017012685.1 534 Missense Mutation GCC,TCC A48S XP_016868174.1
XM_017012686.1 534 Missense Mutation GCC,TCC A48S XP_016868175.1
XM_017012687.1 534 Intron XP_016868176.1
XM_017012688.1 534 Missense Mutation GCC,TCC A48S XP_016868177.1
XM_017012689.1 534 Missense Mutation GCC,TCC A48S XP_016868178.1
Gene
FBXL13
Gene Name
F-box and leucine rich repeat protein 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111038.1 534 Intron NP_001104508.1
NM_001287150.1 534 Intron NP_001274079.1
NM_145032.3 534 Intron NP_659469.3
XM_005250205.3 534 Intron XP_005250262.1
XM_005250207.3 534 Intron XP_005250264.1
XM_005250208.3 534 Intron XP_005250265.1
XM_005250209.2 534 Intron XP_005250266.1
XM_006715898.2 534 Intron XP_006715961.1
XM_011515928.2 534 Intron XP_011514230.1
XM_011515929.2 534 Intron XP_011514231.1
XM_011515930.2 534 Intron XP_011514232.1
XM_011515932.2 534 Intron XP_011514234.1
XM_017011850.1 534 Intron XP_016867339.1
XM_017011851.1 534 Intron XP_016867340.1
XM_017011852.1 534 Intron XP_016867341.1
XM_017011853.1 534 Intron XP_016867342.1

View Full Product Details