Product Details

SNP ID
rs145672737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:550436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGGGCGGGAGCTGTGCTCAGAC[A/G]GTGAGGGAGATGAAGCTGTTGTAAC
Phenotype
MIM: 176911
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRKAR1B PubMed Links

Gene Details

Gene
PRKAR1B
Gene Name
protein kinase cAMP-dependent type I regulatory subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164758.1 1184 Silent Mutation ACC,ACT T380T NP_001158230.1
NM_001164759.1 1184 Silent Mutation ACC,ACT T380T NP_001158231.1
NM_001164760.1 1184 Silent Mutation ACC,ACT T380T NP_001158232.1
NM_001164761.1 1184 Silent Mutation ACC,ACT T380T NP_001158233.1
NM_001164762.1 1184 Silent Mutation ACC,ACT T380T NP_001158234.1
NM_002735.2 1184 Silent Mutation ACC,ACT T380T NP_002726.1
XM_011515446.2 1184 Silent Mutation ACC,ACT T325T XP_011513748.1
XM_017012433.1 1184 Silent Mutation ACC,ACT T250T XP_016867922.1
XM_017012434.1 1184 Silent Mutation ACC,ACT T228T XP_016867923.1

View Full Product Details