Product Details

SNP ID
rs145696649
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:103299622 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGCAGGGAGAGCTCTTTCAGTA[C/G]TATAACATGTTTTATGTTTATTTTT
Phenotype
MIM: 603131
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PMPCB PubMed Links

Gene Details

Gene
PMPCB
Gene Name
peptidase, mitochondrial processing beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004279.2 Intron NP_004270.2
XM_005250717.3 Intron XP_005250774.1
XM_006716181.3 Intron XP_006716244.1

View Full Product Details