Product Details

SNP ID
rs145921800
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73435217 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGCGCTCACGGGCTTCGTGCTG[G/T]TGCCCCTCTCTGGCTACCTGGTGCT
Phenotype
MIM: 605681 MIM: 601766
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BAZ1B PubMed Links

Gene Details

Gene
BAZ1B
Gene Name
bromodomain adjacent to zinc finger domain 1B
There are no transcripts associated with this gene.

Gene
FZD9
Gene Name
frizzled class receptor 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003508.2 1439 Missense Mutation GTG,TTG V404L NP_003499.1

View Full Product Details