Product Details

SNP ID
rs146334064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:2912177 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCCAGTCCCCGCCTCACCTTGG[C/T]GGCCACAGCTTCAATGTTGGCAGGG
Phenotype
MIM: 607210
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CARD11 PubMed Links

Gene Details

Gene
CARD11
Gene Name
caspase recruitment domain family member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324281.1 3543 Missense Mutation NP_001311210.1
NM_032415.5 3543 Missense Mutation NP_115791.3
XM_011515586.2 3543 Missense Mutation XP_011513888.1
XM_011515587.2 3543 Missense Mutation XP_011513889.1

View Full Product Details