Product Details

SNP ID
rs146897803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:54549905 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAGTGAGGAAAAAGGATGAAGG[C/T]TTATATGAGTGCAGGGTGACTGATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VSTM2A PubMed Links

Gene Details

Gene
VSTM2A
Gene Name
V-set and transmembrane domain containing 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301009.1 775 Silent Mutation GGC,GGT G123G NP_001287938.1
NM_001317843.1 775 Silent Mutation GGC,GGT G123G NP_001304772.1
NM_182546.3 775 Silent Mutation GGC,GGT G123G NP_872352.2
XM_006715665.3 775 Silent Mutation GGC,GGT G123G XP_006715728.1
XM_006715666.3 775 Silent Mutation GGC,GGT G123G XP_006715729.1
XM_017011841.1 775 Silent Mutation GGC,GGT G123G XP_016867330.1
Gene
VSTM2A-OT1
Gene Name
VSTM2A overlapping transcript 1
There are no transcripts associated with this gene.

View Full Product Details