Product Details

SNP ID
rs147422861
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:2906649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGGCGCCCGCCTCACAGCTGGT[C/T]CTCGTCCACCCAGATGGTCTTGCGC
Phenotype
MIM: 607210
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CARD11 PubMed Links

Gene Details

Gene
CARD11
Gene Name
caspase recruitment domain family member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324281.1 3858 Missense Mutation AAC,GAC N1152D NP_001311210.1
NM_032415.5 3858 Missense Mutation AAC,GAC N1152D NP_115791.3
XM_011515586.2 3858 Missense Mutation AAC,GAC N1152D XP_011513888.1
XM_011515587.2 3858 Missense Mutation AAC,GAC N1151D XP_011513889.1

View Full Product Details