Product Details

SNP ID
rs147670794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:127611954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCCTCCCGAGTACCTGTGCAGA[A/G]ATGATTCCTGGGGCAACCCTTGGTA
Phenotype
MIM: 167413
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAX4 PubMed Links

Gene Details

Gene
PAX4
Gene Name
paired box 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006193.2 944 Silent Mutation ATC,ATT I246I NP_006184.2
XM_011516276.2 944 Silent Mutation ATC,ATT I254I XP_011514578.1

View Full Product Details