Product Details

SNP ID
rs147870256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27243079 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTTTCTGGATGGGGGTCAGCTT[G/T]GCACTCTGGTTGGCAAGAGAGTCTC
Phenotype
MIM: 142996
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EVX1 PubMed Links

Gene Details

Gene
EVX1
Gene Name
even-skipped homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304519.1 535 UTR 5 NP_001291448.1
NM_001304520.1 535 UTR 5 NP_001291449.1
NM_001989.4 535 Missense Mutation GGC,TGC G17C NP_001980.1
Gene
EVX1-AS
Gene Name
EVX1 antisense RNA
There are no transcripts associated with this gene.

View Full Product Details