Product Details

SNP ID
rs148341129
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6466167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTCAAAGAAGCCCTCAAAGTAGA[A/C]GCGCCAGATCCAGTTGACAAGATAC
Phenotype
MIM: 609024
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KDELR2 PubMed Links

Gene Details

Gene
KDELR2
Gene Name
KDEL endoplasmic reticulum protein retention receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100603.1 669 Intron NP_001094073.1
NM_006854.3 669 Missense Mutation GTC,TTC V170F NP_006845.1

View Full Product Details