Product Details

SNP ID
rs149259090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:103299475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGATTGATGCTGGAAGTAGATA[C/T]GAAAATGAGAAGAACAATGGAACAG
Phenotype
MIM: 603131
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PMPCB PubMed Links

Gene Details

Gene
PMPCB
Gene Name
peptidase, mitochondrial processing beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004279.2 307 Silent Mutation TAC,TAT Y91Y NP_004270.2
XM_005250717.3 307 Nonsense Mutation TAC,TAT Y91Y XP_005250774.1
XM_006716181.3 307 Nonsense Mutation TAC,TAT Y91Y XP_006716244.1

View Full Product Details