Product Details

SNP ID
rs149439578
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44827520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCTGAAGAATCTGAGCCAAACT[A/G]GAAGAAACCAGCCTCATTTCCAGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
H2AFV PubMed Links

Gene Details

Gene
H2AFV
Gene Name
H2A histone family member V
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012412.4 3124 Intron NP_036544.1
NM_138635.3 3124 UTR 3 NP_619541.1
NM_201436.2 3124 Intron NP_958844.1
NM_201516.2 3124 Intron NP_958924.1
NM_201517.2 3124 Intron NP_958925.1

View Full Product Details