Product Details

SNP ID
rs149872164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99928908 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCGGTTGCTTTCTTGTGTCTTCTG[A/G]TGCTCTCTGAAGTTAGGAAGTATTT
Phenotype
MIM: 611925
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GJC3 PubMed Links

Gene Details

Gene
GJC3
Gene Name
gap junction protein gamma 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181538.2 713 Missense Mutation ACC,ATC T238I NP_853516.1
Gene
LOC101927610
Gene Name
uncharacterized LOC101927610
There are no transcripts associated with this gene.

Gene
TRIM4
Gene Name
tripartite motif containing 4
There are no transcripts associated with this gene.

View Full Product Details