Product Details
- SNP ID
-
rs150224436
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:101616233 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGGAAACTTACACATCGGCCTTGA[C/T]GAAACCTTTCCCTTCAGTGTCGAAC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MYL10
PubMed Links
Gene Details
- Gene
- MYL10
- Gene Name
- myosin light chain 10
View Full Product Details