Product Details

SNP ID
rs151311760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:122304153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGGGCCTCCAGACTGGCCTTCCG[C/G]GGCTCGGAGCCCGTCACTCCTGCCT
Phenotype
MIM: 613301
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FEZF1 PubMed Links

Gene Details

Gene
FEZF1
Gene Name
FEZ family zinc finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024613.3 2030 Silent Mutation CCC,CCG P95P NP_001019784.2
NM_001160264.2 2030 Silent Mutation CCC,CCG P95P NP_001153736.1
XM_005250337.3 2030 Silent Mutation CCC,CCG P95P XP_005250394.1
XM_011516202.2 2030 Silent Mutation CCC,CCG P95P XP_011514504.1
Gene
FEZF1-AS1
Gene Name
FEZF1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details