Product Details

SNP ID
rs111267981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:43300535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTACAGGGTGGCCTCACACCACT[C/T]TTGCTTGCTGTACATGGACAAAAAC
Phenotype
MIM: 608915
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POTEA PubMed Links

Gene Details

Gene
POTEA
Gene Name
POTE ankyrin domain family member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002920.1 Intron NP_001002920.1
NM_001005365.2 Intron NP_001005365.2
XM_011544516.1 Intron XP_011542818.1

View Full Product Details