Product Details

SNP ID
rs112204704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:96273336 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGAAGACAACATCTGGAGAGGC[A/G]TCCTCTCTGTTATTTTCTTCTTTCT
Phenotype
MIM: 612792
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTDSS1 PubMed Links

Gene Details

Gene
PTDSS1
Gene Name
phosphatidylserine synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290225.1 372 UTR 5 NP_001277154.1
NM_014754.2 372 Missense Mutation ATC,GTC I73V NP_055569.1

View Full Product Details