Product Details

SNP ID
rs116451664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:6925898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCAAAGGAGACGGCAAAGTCCTG[C/G]TCATTTGCCCCACGCTGCTCCTGGG
Phenotype
MIM: 601157 MIM: 600471
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DEFA4 PubMed Links

Gene Details

Gene
DEFA4
Gene Name
defensin alpha 4
There are no transcripts associated with this gene.

Gene
DEFA6
Gene Name
defensin alpha 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001926.3 179 Missense Mutation GAC,GAG D46E NP_001917.1

View Full Product Details