Product Details

SNP ID
rs116938239
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:25844672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAAGAGTGGCACAGGAATGAGAC[C/T]TGGGGACTTTTTATGTTCAAGGCTA
Phenotype
MIM: 609934
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EBF2 PubMed Links

Gene Details

Gene
EBF2
Gene Name
early B-cell factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022659.3 Intron NP_073150.2

View Full Product Details