Product Details

SNP ID
rs139248084
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:66494069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGATAGAGACTTTCATTTTTTCCA[G/T]TGAGCAAGTGGGTTCTGAAATGAGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C8orf46 PubMed Links

Gene Details

Gene
C8orf46
Gene Name
chromosome 8 open reading frame 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152765.3 Intron NP_689978.2

View Full Product Details