Product Details

SNP ID
rs139449787
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:91219025 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACAGGTGAGGTGCTCATTCATAG[A/C]CTCACTCCACTTGCCCTGTCAGCTC
Phenotype
MIM: 608479
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LRRC69 PubMed Links

Gene Details

Gene
LRRC69
Gene Name
leucine rich repeat containing 69
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129890.1 1112 UTR 3 NP_001123362.1
Gene
SLC26A7
Gene Name
solute carrier family 26 member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282356.1 1112 Intron NP_001269285.1
NM_001282357.1 1112 Intron NP_001269286.1
NM_052832.3 1112 Intron NP_439897.1
NM_134266.1 1112 Intron NP_599028.1

View Full Product Details