Product Details

SNP ID
rs139543288
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22583331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGAAAGGGATCAGGAGTGACGC[A/G]CTCAAAGGCACCCTGGCCTCCAGTG
Phenotype
MIM: 609722 MIM: 610795
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDLIM2 PubMed Links

Gene Details

Gene
PDLIM2
Gene Name
PDZ and LIM domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021630.5 Intron NP_067643.3
NM_176871.3 Intron NP_789847.1
NM_198042.3 Intron NP_932159.1
Gene
SORBS3
Gene Name
sorbin and SH3 domain containing 3
There are no transcripts associated with this gene.

View Full Product Details