Product Details

SNP ID
rs139620518
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143614210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCGGATTCCCTGTACCCCACACC[A/G]TCAGGGCCGAGCCGCTGCCTGCAGA
Phenotype
MIM: 616408 MIM: 137020
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PYCRL PubMed Links

Gene Details

Gene
PYCRL
Gene Name
pyrroline-5-carboxylate reductase-like
There are no transcripts associated with this gene.

Gene
TSTA3
Gene Name
tissue specific transplantation antigen P35B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317783.1 744 Missense Mutation ACG,ATG T212M NP_001304712.1
NM_003313.3 744 Missense Mutation ACG,ATG T206M NP_003304.1
XM_005251051.3 744 Missense Mutation ACG,ATG T206M XP_005251108.2
XM_011517269.1 744 Missense Mutation ACG,ATG T206M XP_011515571.1

View Full Product Details