Product Details

SNP ID
rs139926450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:9556224 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCTGTTGCAGTACCACCAGCAC[A/T]ATCTGTACCGTCGCCGCCGCTCCCG
Phenotype
MIM: 603303
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TNKS PubMed Links

Gene Details

Gene
TNKS
Gene Name
tankyrase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003747.2 996 Silent Mutation ACA,ACT T95T NP_003738.2
XM_006716263.3 996 Silent Mutation ACA,ACT T95T XP_006716326.1
XM_011543845.2 996 Silent Mutation ACA,ACT T95T XP_011542147.1
XM_011543846.2 996 Silent Mutation ACA,ACT T95T XP_011542148.1
XM_011543847.2 996 Silent Mutation ACA,ACT T95T XP_011542149.1

View Full Product Details