Product Details

SNP ID
rs140337555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142613525 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGAACTCCCACCACTTCTTGGC[C/T]GGCCCGTTCATGTGATTCTGGATCT
Phenotype
MIM: 612461
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARC PubMed Links

Gene Details

Gene
ARC
Gene Name
activity regulated cytoskeleton associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015193.4 948 Silent Mutation CCA,CCG P249P NP_056008.1

View Full Product Details