Product Details

SNP ID
rs141207599
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:86518518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACTGATTTTCATTGAGGGTAAAC[A/G]GTGATTCACTTTGTAGACATGTATT
Phenotype
MIM: 604207 MIM: 611871
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CPNE3 PubMed Links

Gene Details

Gene
CPNE3
Gene Name
copine 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003909.4 Intron NP_003900.1
XM_005251093.4 Intron XP_005251150.1
XM_017013945.1 Intron XP_016869434.1
Gene
RMDN1
Gene Name
regulator of microtubule dynamics 1
There are no transcripts associated with this gene.

View Full Product Details