Product Details

SNP ID
rs142518057
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143044697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGGACTCACTGCCAAAGACGCT[C/T]ACTTTCTGGGAGACACAGACCCCAT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
C8orf31 PubMed Links

Gene Details

Gene
C8orf31
Gene Name
chromosome 8 open reading frame 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173687.3 902 Missense Mutation CAC,TAC H79Y NP_775958.1
XM_017013344.1 902 Missense Mutation CAC,TAC H91Y XP_016868833.1
XM_017013345.1 902 Missense Mutation CAC,TAC H91Y XP_016868834.1
XM_017013346.1 902 Missense Mutation CAC,TAC H74Y XP_016868835.1
Gene
LOC107984097
Gene Name
uncharacterized protein FLJ40521
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017014161.1 902 Intron XP_016869650.1

View Full Product Details