Product Details

SNP ID
rs143344126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42708832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACATTCTAATGACACCATAAAAGT[A/G]TATTTTGGATTGAAAATATCCCAGC
Phenotype
MIM: 118508
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHRNB3 PubMed Links

Gene Details

Gene
CHRNB3
Gene Name
cholinergic receptor nicotinic beta 3 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000749.3 296 Silent Mutation GTA,GTG V56V NP_000740.1
XM_011544390.1 296 Intron XP_011542692.1

View Full Product Details