Product Details

SNP ID
rs143450315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:10054578 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCCACAGCCTCTTTCCCGTCC[C/G]GAGGATGGGCAACTCGGCCTCGAAC
Phenotype
MIM: 601250
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MSRA PubMed Links

Gene Details

Gene
MSRA
Gene Name
methionine sulfoxide reductase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135670.2 310 Missense Mutation CCG,CGG P21R NP_001129142.1
NM_001135671.2 310 Intron NP_001129143.1
NM_001199729.2 310 Intron NP_001186658.1
NM_012331.4 310 Missense Mutation CCG,CGG P21R NP_036463.1
XM_011543822.1 310 Missense Mutation CCG,CGG P21R XP_011542124.1
XM_011543823.2 310 Missense Mutation CCG,CGG P21R XP_011542125.1
XM_017013448.1 310 Missense Mutation CCG,CGG P21R XP_016868937.1
XM_017013449.1 310 Intron XP_016868938.1
XM_017013450.1 310 Intron XP_016868939.1
XM_017013451.1 310 Intron XP_016868940.1
XM_017013452.1 310 Intron XP_016868941.1
XM_017013453.1 310 Intron XP_016868942.1
XM_017013454.1 310 Intron XP_016868943.1

View Full Product Details