Product Details

SNP ID
rs145192530
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:33500394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTGTGGCCTCCTGTAGCAGGGC[A/G]CTCTTAACAGACTCAGGTGTAAGGT
Phenotype
MIM: 614426
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAK16 PubMed Links

Gene Details

Gene
MAK16
Gene Name
MAK16 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032509.3 1882 UTR 3 NP_115898.2
Gene
TTI2
Gene Name
TELO2 interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102401.2 1882 Silent Mutation AGC,AGT S452S NP_001095871.1
NM_001265581.1 1882 Silent Mutation AGC,AGT S452S NP_001252510.1
NM_025115.3 1882 Silent Mutation AGC,AGT S452S NP_079391.2
XM_011544665.2 1882 Silent Mutation AGC,AGT S421S XP_011542967.1

View Full Product Details