Product Details

SNP ID
rs145997578
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:24294097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGAGGAGGCAGGGACAGACCCAG[C/T]AGCACCCACCTGAGCGAGAAGAGCA
Phenotype
MIM: 606188
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAM28 PubMed Links

Gene Details

Gene
ADAM28
Gene Name
ADAM metallopeptidase domain 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304351.1 58 UTR 5 NP_001291280.1
NM_014265.5 58 UTR 5 NP_055080.2
NM_021777.4 58 UTR 5 NP_068547.2
XM_005273380.3 58 UTR 5 XP_005273437.1
XM_005273382.1 58 UTR 5 XP_005273439.1
XM_006716273.3 58 UTR 5 XP_006716336.1
XM_006716274.1 58 UTR 5 XP_006716337.1
XM_011544367.2 58 UTR 5 XP_011542669.1
XM_011544368.2 58 UTR 5 XP_011542670.1
XM_011544369.2 58 UTR 5 XP_011542671.1
XM_011544370.2 58 UTR 5 XP_011542672.1
XM_011544371.2 58 Intron XP_011542673.1
XM_017012974.1 58 UTR 5 XP_016868463.1
XM_017012975.1 58 UTR 5 XP_016868464.1
XM_017012976.1 58 UTR 5 XP_016868465.1
Gene
LOC101929294
Gene Name
uncharacterized LOC101929294
There are no transcripts associated with this gene.

View Full Product Details