Product Details

SNP ID
rs146359054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:62979480 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTACCCTTTTCTAAATAATCAC[A/G]TTGCTAAAATGTGCCACATGGCCCA
Phenotype
MIM: 612872
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NKAIN3 PubMed Links

Gene Details

Gene
NKAIN3
Gene Name
Na+/K+ transporting ATPase interacting 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304533.1 Intron NP_001291462.1
XM_011517511.2 Intron XP_011515813.1
XM_011517512.2 Intron XP_011515814.1
XM_017013359.1 Intron XP_016868848.1
XM_017013360.1 Intron XP_016868849.1
XM_017013361.1 Intron XP_016868850.1
Gene
UG0898H09
Gene Name
uncharacterized LOC643763
There are no transcripts associated with this gene.

View Full Product Details