Product Details

SNP ID
rs146601365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:491874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATAAGCTTTGCTTTTCCAGTAT[C/T]TGTTTACGTATTTGTTTAATAAGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TDRP PubMed Links

Gene Details

Gene
TDRP
Gene Name
testis development related protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256113.1 1653 Intron NP_001243042.1
NM_175075.4 1653 UTR 3 NP_778250.2
XM_006716232.3 1653 UTR 3 XP_006716295.1
XM_017013113.1 1653 UTR 3 XP_016868602.1

View Full Product Details