Product Details
- SNP ID
-
rs147143248
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:107253264 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATGTTAATGTAGTCAAAACATGAG[G/T]GACAAAAAGGTTCTTGTTTGGAAAA
- Phenotype
-
MIM: 601667
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ANGPT1
PubMed Links
Gene Details
- Gene
- ANGPT1
- Gene Name
- angiopoietin 1
View Full Product Details