Product Details

SNP ID
rs147680673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:52539838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACCCAAGTTACTTACAAGCTGG[C/T]GTTGAGCACTCCCTGGTATTGTAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM150A PubMed Links

Gene Details

Gene
FAM150A
Gene Name
family with sequence similarity 150 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207413.3 523 Silent Mutation ACA,ACG T106T NP_997296.1

View Full Product Details